Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749899.3 | XP_003749947.1 | 33 | 7357 | ||
XM_006237848.2 | XP_006237910.1 | 33 | 7354 |