Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DGKD SIRNA Assays ›
Gene Symbol:
DGKD
Gene Name:
diacylglycerol kinase delta
Gene Aliases:
DGKdelta, dgkd-2
Chromosome Location:
Chr.2: 233354507 - 233472098 on Build GRCh38
UniGene ID:
Hs.471675
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16209
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DGKD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_003648.2
NP_003639.2
17, 18
2216
NM_152879.2
NP_690618.2
18, 19
2281
XM_005246107.3
XP_005246164.1
16, 17
1948
XM_005246108.1
XP_005246165.1
16, 17
2102
XM_005246110.3
XP_005246167.1
7, 8
1953
XM_011512028.2
XP_011510330.1
18, 19
3740
XM_011512029.2
XP_011510331.1
18, 19
3741
XM_011512030.2
XP_011510332.1
18, 19
2337
XM_011512031.2
XP_011510333.1
18, 19
2211
XM_011512032.2
XP_011510334.1
18, 19
2195
XM_011512033.2
XP_011510335.1
18, 19
3742
XM_011512035.1
XP_011510337.1
16, 17
2082
XM_011512037.2
XP_011510339.1
16, 17
2186
XM_011512039.2
XP_011510341.1
18, 19
3742
XM_011512040.2
XP_011510342.1
16, 17
2118
XM_017005138.1
XP_016860627.1
18, 19
2619
XM_017005139.1
XP_016860628.1
16, 17
2111
XM_017005140.1
XP_016860629.1
18, 19
2213
GenBank mRNA
AB078966.1
-
18, 19
2281
AK307426.1
-
6, 7
839
AK308846.1
-
16, 17
2102
AK311044.1
-
16, 17
2035
D63479.3
-
18, 19
2222
D73409.1
-
17, 18
2217
Back To Top
More Information
Panther Classification:
Molecular Function -
kinase
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
signal transduction
epidermal growth factor receptor signaling pathway
protein kinase C-activating G-protein coupled receptor signaling pathway
multicellular organism development
response to organic substance
protein transport
cell growth
phosphorylation
second-messenger-mediated signaling
platelet activation
diacylglycerol metabolic process
protein homooligomerization
diacylglycerol kinase activity
protein binding
ATP binding
diacylglycerol binding
protein homodimerization activity
metal ion binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0