Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004312.2 | NP_004303.2 | 5, 6 | 212 | ||
XM_017029518.1 | XP_016885007.1 | 3, 4 | 189 | |||
GenBank mRNA | AF033105.1 | - | 5, 6 | 195 | ||
BC012096.1 | - | 5, 6 | 242 | 4645867 | ||
BG472156.1 | - | 5, 6 | 243 | |||
CR456849.1 | - | 3, 4 | 144 | |||
EU883571.1 | - | 3, 4 | 144 | |||
U03626.1 | - | 3, 4 | 144 |