Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003648.2 | NP_003639.2 | 8, 9 | 1039 | ||
NM_152879.2 | NP_690618.2 | 9, 10 | 1104 | |||
XM_005246107.3 | XP_005246164.1 | 7, 8 | 771 | |||
XM_005246108.1 | XP_005246165.1 | 7, 8 | 925 | |||
XM_011512028.2 | XP_011510330.1 | 9, 10 | 2563 | |||
XM_011512029.2 | XP_011510331.1 | 9, 10 | 2564 | |||
XM_011512030.2 | XP_011510332.1 | 9, 10 | 1160 | |||
XM_011512031.2 | XP_011510333.1 | 9, 10 | 1034 | |||
XM_011512032.2 | XP_011510334.1 | 9, 10 | 1018 | |||
XM_011512033.2 | XP_011510335.1 | 9, 10 | 2565 | |||
XM_011512035.1 | XP_011510337.1 | 7, 8 | 905 | |||
XM_011512037.2 | XP_011510339.1 | 7, 8 | 1009 | |||
XM_011512039.2 | XP_011510341.1 | 9, 10 | 2565 | |||
XM_011512040.2 | XP_011510342.1 | 7, 8 | 941 | |||
XM_017005138.1 | XP_016860627.1 | 9, 10 | 1442 | |||
XM_017005139.1 | XP_016860628.1 | 7, 8 | 934 | |||
XM_017005140.1 | XP_016860629.1 | 9, 10 | 1036 | |||
GenBank mRNA | AB078966.1 | - | 9, 10 | 1104 | ||
AK308846.1 | - | 7, 8 | 925 | |||
AK311044.1 | - | 7, 8 | 858 | |||
D63479.3 | - | 9, 10 | 1045 | |||
D73409.1 | - | 8, 9 | 1040 |