Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173507.1 | NP_001166978.1 | 29 | 3582 | ||
XM_006257503.2 | XP_006257565.1 | 29 | 4618 | |||
XM_008773521.1 | XP_008771743.1 | 29 | 3752 | |||
XM_008773522.1 | XP_008771744.1 | 29 | 3316 | |||
XM_008773523.1 | XP_008771745.1 | 29 | 4275 |