Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175765.2 | NP_786941.1 | 11 | 1163 | ||
XM_006238353.2 | XP_006238415.1 | 12 | 1390 | |||
XM_006238354.2 | XP_006238416.1 | 12 | 1386 | |||
GenBank mRNA | AB285525.1 | - | 1066 | |||
AF339084.1 | - | 11 | 1163 |