Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238127.2 | XP_006238189.1 | 7 | 2449 | ||
XM_233141.8 | XP_233141.7 | 7 | 2992 |