Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131012.1 | NP_001124484.1 | 12 | 1194 | ||
XM_006238479.1 | XP_006238541.1 | 13 | 1553 | |||
XM_006238483.2 | XP_006238545.1 | 14 | 1310 | |||
XM_006238484.2 | XP_006238546.1 | 14 | 1420 | |||
XM_008763874.1 | XP_008762096.1 | 12 | 4836 | |||
XM_008763875.1 | XP_008762097.1 | 13 | 4277 | |||
XM_008763876.1 | XP_008762098.1 | 13 | 4108 | |||
XM_008763877.1 | XP_008762099.1 | 13 | 4752 | |||
XM_008763878.1 | XP_008762100.1 | 13 | 2276 | |||
XM_008763879.1 | XP_008762101.1 | 13 | 2273 | |||
XM_008763880.1 | XP_008762102.1 | 10 | 1093 | |||
XM_008763881.1 | XP_008762103.1 | 13 | 1470 | |||
GenBank mRNA | BC168252.1 | - | 12 | 1194 | 5600058 |