Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107966.1 | NP_001101436.1 | 16 | 2397 | ||
XM_006238668.1 | XP_006238730.1 | 17 | 2605 | |||
XM_006238670.2 | XP_006238732.1 | 15 | 2292 |