Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077671.1 | NP_001071139.1 | 2 | 256 | ||
XM_006238865.2 | XP_006238927.1 | 1 | 886 | |||
GenBank mRNA | BC127516.1 | - | 2 | 256 | 7102954 |