Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107986.2 | NP_001101456.2 | 2 | 118 | ||
XM_006239182.2 | XP_006239244.1 | 2 | 205 | |||
XM_006239183.2 | XP_006239245.1 | 2 | 205 | |||
XM_006239184.2 | XP_006239246.1 | 2 | 206 | |||
XM_006239185.2 | XP_006239247.1 | 2 | 205 | |||
XM_008764242.1 | XP_008762464.1 | 2 | 335 | |||
XM_008764243.1 | XP_008762465.1 | 2 | 336 | |||
GenBank mRNA | BC166715.1 | - | 2 | 118 | 7930852 |