Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191770.1 | NP_001178699.1 | 25 | 3868 | ||
XM_008764459.1 | XP_008762681.1 | 26 | 4297 | |||
XM_008764460.1 | XP_008762682.1 | 26 | 4005 | |||
XM_008764461.1 | XP_008762683.1 | 26 | 3997 | |||
XM_008764462.1 | XP_008762684.1 | 25 | 4376 | |||
XM_008764463.1 | XP_008762685.1 | 24 | 3802 |