Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320687.1 | NP_001307616.1 | 11 | 1993 | ||
NM_016166.2 | NP_057250.1 | 10 | 1394 | |||
XM_011522126.2 | XP_011520428.1 | 12 | 2101 | |||
XM_011522127.2 | XP_011520429.1 | 10 | 1878 | |||
XM_017022688.1 | XP_016878177.1 | 10 | 1872 | |||
XM_017022689.1 | XP_016878178.1 | 10 | 1885 | |||
XM_017022690.1 | XP_016878179.1 | 10 | 1690 | |||
XM_017022691.1 | XP_016878180.1 | 10 | 1464 | |||
GenBank mRNA | AF077951.1 | - | 10 | 1243 | ||
AF167160.1 | - | 10 | 1342 | |||
AK094641.1 | - | 11 | 1993 | |||
AK314515.1 | - | 10 | 1269 | |||
AY826819.1 | - | 10 | 1243 | |||
BC053685.1 | - | 10 | 1247 | |||
BC118587.1 | - | 10 | 1296 | 40085010 | ||
BC121797.1 | - | 10 | 1293 | 40085007 | ||
EF487997.1 | - | 10 | 1246 | |||
EF487998.1 | - | 10 | 1246 | |||
U78524.1 | - | 9 | 1228 |