Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108012.1 | NP_001101482.1 | 12 | 2248 | ||
XM_006239842.2 | XP_006239904.1 | 10 | 3328 | |||
XM_006239843.2 | XP_006239905.1 | 10 | 2560 | |||
XM_006239844.2 | XP_006239906.1 | 10 | 3328 | |||
XM_006239845.2 | XP_006239907.1 | 10 | 3328 | |||
XM_006239846.2 | XP_006239908.1 | 10 | 3328 |