Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankmy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108019.1 | NP_001101489.1 | 2 | 286 | ||
XM_006240042.2 | XP_006240104.1 | 2 | 2671 | |||
XM_006240043.2 | XP_006240105.2 | 3 | 2988 |