Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108029.1 | NP_001101499.1 | 7 | 732 | ||
GenBank mRNA | FQ233288.1 | - | 9 | 809 |