Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108043.1 | NP_001101513.1 | 2 | 346 | ||
XM_006240310.2 | XP_006240372.1 | 2 | 1007 | |||
XM_006240311.2 | XP_006240373.1 | 2 | 711 | |||
XM_006240312.2 | XP_006240374.1 | 2 | 711 | |||
XM_006240313.2 | XP_006240375.1 | 2 | 708 | |||
XM_006240314.2 | XP_006240376.1 | 2 | 708 |