Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MADD SIRNA Assays ›
Gene Symbol:
MADD
Gene Name:
MAP kinase activating death domain
Gene Aliases:
DENN, IG20, RAB3GEP
Chromosome Location:
Chr.11: 47269376 - 47330031 on Build GRCh38
UniGene ID:
Hs.82548
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16315
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MADD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135943.1
NP_001129415.1
18
3169
NM_001135944.1
NP_001129416.1
18
3145
NM_003682.3
NP_003673.3
19
3334
NM_130470.2
NP_569826.2
18
3440
NM_130471.2
NP_569827.2
19
3205
NM_130472.2
NP_569828.2
18
3145
NM_130473.2
NP_569829.2
19
3334
NM_130474.2
NP_569830.2
18
3145
NM_130475.2
NP_569831.1
19
3358
NM_130476.2
NP_569832.2
18
3298
XM_005253189.2
XP_005253246.1
19
3358
XM_005253196.2
XP_005253253.1
19
3358
XM_005253199.2
XP_005253256.1
18
3298
XM_005253200.2
XP_005253257.1
19
3358
XM_005253201.2
XP_005253258.1
19
3229
XM_005253203.2
XP_005253260.1
18
3169
XM_005253204.2
XP_005253261.1
18
3169
XM_005253205.1
XP_005253262.1
18
3169
XM_017018478.1
XP_016873967.1
19
3358
XM_017018479.1
XP_016873968.1
19
3358
XM_017018480.1
XP_016873969.1
19
3358
XM_017018481.1
XP_016873970.1
18
3298
XM_017018482.1
XP_016873971.1
19
3358
XM_017018483.1
XP_016873972.1
19
3229
XM_017018484.1
XP_016873973.1
18
3298
XM_017018485.1
XP_016873974.1
18
3259
XM_017018486.1
XP_016873975.1
19
3229
XM_017018487.1
XP_016873976.1
18
3169
XM_017018488.1
XP_016873977.1
19
3358
XM_017018489.1
XP_016873978.1
19
3319
XM_017018490.1
XP_016873979.1
19
3345
XM_017018491.1
XP_016873980.1
19
3314
XM_017018492.1
XP_016873981.1
19
3358
XM_017018493.1
XP_016873982.1
18
3285
XM_017018494.1
XP_016873983.1
18
3298
XM_017018495.1
XP_016873984.1
18
3298
XM_017018496.1
XP_016873985.1
18
3254
XM_017018497.1
XP_016873986.1
18
3259
XM_017018498.1
XP_016873987.1
19
3229
XM_017018499.1
XP_016873988.1
19
3198
XM_017018500.1
XP_016873989.1
18
3298
XM_017018501.1
XP_016873990.1
19
3185
XM_017018502.1
XP_016873991.1
19
3198
XM_017018503.1
XP_016873992.1
19
3185
XM_017018504.1
XP_016873993.1
19
3198
XM_017018505.1
XP_016873994.1
19
3185
XM_017018506.1
XP_016873995.1
19
3190
XM_017018507.1
XP_016873996.1
18
3156
XM_017018508.1
XP_016873997.1
18
3169
XM_017018509.1
XP_016873998.1
18
3125
XM_017018510.1
XP_016873999.1
18
3169
GenBank mRNA
AB002356.1
-
19
3330
AF440100.1
-
19
3308
AF440101.1
-
19
3308
AF440102.1
-
19
3179
AF440103.1
-
18
3119
AF440434.1
-
18
3119
AK292442.1
-
18
3109
BC040484.1
-
18
3153
5271798
U44953.1
-
18
3262
U77352.1
-
18
3408
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001177719, NM_145527, NM_001177720, AK036347, AY263984, and others.
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
cell surface receptor signaling pathway
regulation of tumor necrosis factor-mediated signaling pathway
regulation of Rab protein signal transduction
regulation of apoptotic process
positive regulation of GTPase activity
regulation of cell cycle
execution phase of apoptosis
regulation of extrinsic apoptotic signaling pathway via death domain receptors
regulation of extrinsic apoptotic signaling pathway
death receptor binding
protein binding
Rab guanyl-nucleotide exchange factor activity
protein kinase activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0