Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR7A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320979.1 | NP_001307908.1 | 4 | 682 | ||
NM_003689.3 | NP_003680.2 | 5 | 787 | |||
GenBank mRNA | AF026947.1 | - | 5 | 755 | ||
BC004111.2 | - | 5 | 714 | 3832140 | ||
BC007352.2 | - | 5 | 762 | |||
BC010852.1 | - | 5 | 763 | 3898226 | ||
BC011586.1 | - | 5 | 714 | 4620484 | ||
BC012171.1 | - | 5 | 762 | 4642811 | ||
BC013996.2 | - | 5 | 718 | 3625337 | ||
BT007347.1 | - | 5 | 678 | |||
BX401279.2 | - | 4 | 630 | |||
EU794591.1 | - | 5 | 787 | |||
Y16675.1 | - | 5 | 749 |