Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039027.1 | NP_001034116.1 | 2 | 267 | ||
XM_006240906.1 | XP_006240968.1 | 2 | 344 | |||
GenBank mRNA | BC099828.1 | - | 2 | 267 | 7934398 | |
FQ212635.1 | - | 2 | 270 | |||
FQ212816.1 | - | 2 | 270 | |||
FQ219525.1 | - | 2 | 270 |