Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dohh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025006.1 | NP_001020177.1 | 5 | 838 | ||
XM_006240937.2 | XP_006240999.1 | 5 | 940 | |||
XM_006240938.2 | XP_006241000.1 | 6 | 1130 | |||
XM_006240939.2 | XP_006241001.1 | 6 | 1127 | |||
GenBank mRNA | BC087658.1 | - | 5 | 838 | 7314221 | |
FQ228684.1 | - | 5 | 904 |