Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008910.2 | NP_001008910.1 | 7 | 918 | ||
NR_026909.1 | 7 | 1183 | ||||
XM_011512067.2 | XP_011510369.1 | 6 | 823 | |||
XM_017005160.1 | XP_016860649.1 | 6 | 1018 | |||
GenBank mRNA | AB020739.1 | - | 6 | 812 | ||
AF060798.1 | - | 7 | 889 | |||
AF203910.1 | - | 7 | 885 | |||
AJ005791.1 | - | 6 | 879 | |||
AJ010872.1 | - | 6 | 789 | |||
AK096954.1 | - | 5 | 1592 | |||
AK225392.1 | - | 7 | 883 | |||
AK225526.1 | - | 864 | ||||
AK292694.1 | - | 7 | 918 | |||
AK298471.1 | - | 5 | 552 | |||
BC002618.2 | - | 7 | 887 | 3143105 | ||
BC053998.1 | - | 7 | 883 | 5760866 | ||
BT019585.1 | - | 6 | 761 | |||
CR407675.1 | - | 6 | 761 |