Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013172.1 | NP_001013190.1 | 1 | 728 | ||
XM_006241179.2 | XP_006241241.1 | 2 | 1087 | |||
GenBank mRNA | BC078961.1 | - | 1 | 728 | 7111014 | |
BC099213.1 | - | 2 | 1061 | 7384591 |