Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003693.3 | NP_003684.2 | 11 | 2907 | ||
NM_145350.2 | NP_663325.1 | 11 | 2870 | |||
NR_028075.2 | 11 | 2837 | ||||
NR_102409.1 | 11 | 2944 | ||||
XM_017025256.1 | XP_016880745.1 | 7 | 1808 | |||
GenBank mRNA | AB052947.1 | - | 11 | 2771 | ||
AB052948.1 | - | 11 | 2837 | |||
AB052949.1 | - | 10 | 3246 | |||
AB052950.1 | - | 9 | 2573 | |||
BC039735.1 | - | 11 | 2875 | 6041360 | ||
D63483.1 | - | 11 | 2817 | |||
D86864.1 | - | 11 | 2841 |