Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271371.1 | NP_001258300.1 | 20 | 3167 | ||
XM_006241248.2 | XP_006241310.1 | 7 | 1285 | |||
XM_006241249.2 | XP_006241311.1 | 5 | 1032 | |||
XM_008765239.1 | XP_008763461.1 | 8 | 1548 | |||
XM_008765240.1 | XP_008763462.1 | 7 | 1288 | |||
XM_008765241.1 | XP_008763463.1 | 7 | 1369 | |||
XM_008765243.1 | XP_008763465.1 | 6 | 1213 | |||
GenBank mRNA | DY315019.1 | - | 3 | 442 |