Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012121.1 | NP_001012121.1 | 4 | 473 | ||
XM_008765722.1 | XP_008763944.1 | 5 | 1569 | |||
XM_008765723.1 | XP_008763945.1 | 4 | 329 | |||
XM_008765724.1 | XP_008763946.1 | 4 | 1154 | |||
XM_008765725.1 | XP_008763947.1 | 5 | 306 | |||
XM_008765726.1 | XP_008763948.1 | 5 | 301 | |||
GenBank mRNA | BC090007.1 | - | 4 | 473 | 7323849 |