Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127547.1 | NP_001121019.1 | 8 | 963 | ||
XM_006242161.2 | XP_006242223.1 | 8 | 943 | |||
GenBank mRNA | BC161973.1 | - | 8 | 925 | 9029287 |