Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108106.1 | NP_001101576.1 | 14 | 2658 | ||
XM_006242202.2 | XP_006242264.1 | 14 | 2428 | |||
XM_006242205.2 | XP_006242267.1 | 15 | 4896 | |||
XM_006242206.2 | XP_006242268.1 | 16 | 2643 | |||
XM_006242207.2 | XP_006242269.1 | 15 | 2551 | |||
XM_008765727.1 | XP_008763949.1 | 14 | 2428 | |||
XM_008765728.1 | XP_008763950.1 | 14 | 4734 | |||
XM_008765729.1 | XP_008763951.1 | 13 | 2378 |