Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134714.1 | NP_001128186.1 | 3 | 664 | ||
XM_006242341.2 | XP_006242403.1 | 4 | 505 | |||
GenBank mRNA | BC168176.1 | - | 3 | 664 | 9034655 |