Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckap1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108119.1 | NP_001101589.1 | 14 | 1423 | ||
XM_008765753.1 | XP_008763975.1 | 14 | 1479 |