Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003718.4 | NP_003709.3 | 2 | 1568 | ||
NM_031267.3 | NP_112557.2 | 2 | 1568 | |||
XM_011515597.2 | XP_011513899.1 | 2 | 2427 | |||
XM_017012750.1 | XP_016868239.1 | 2 | 2427 | |||
XM_017012751.1 | XP_016868240.1 | 2 | 2427 | |||
GenBank mRNA | AJ297709.1 | - | 2 | 1418 | ||
AJ297710.1 | - | 2 | 1418 | |||
AK092534.1 | - | 2 | 1568 | |||
BC080601.1 | - | 2 | 1891 | |||
M80629.1 | - | 2 | 215 |