Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000047.2 | NP_000038.2 | 6 | 702 | ||
NM_001282628.1 | NP_001269557.1 | 7 | 988 | |||
NM_001282631.1 | NP_001269560.1 | 5 | 472 | |||
XM_005274518.2 | XP_005274575.1 | 6 | 629 | |||
XM_005274519.4 | XP_005274576.1 | 6 | 792 | |||
XM_005274521.4 | XP_005274578.1 | 5 | 585 | |||
XM_017029525.1 | XP_016885014.1 | 7 | 919 | |||
GenBank mRNA | AK223183.1 | - | 6 | 532 | ||
AK223199.1 | - | 6 | 499 | |||
AK293648.1 | - | 5 | 407 | |||
AK301004.1 | - | 7 | 819 | |||
AK313093.1 | - | 6 | 521 | |||
BC130438.1 | - | 6 | 497 | 40146469 | ||
X83573.1 | - | 6 | 519 |