Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASET2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003730.4 | NP_003721.2 | 9 | 997 | ||
XM_017011397.1 | XP_016866886.1 | 10 | 776 | |||
XM_017011398.1 | XP_016866887.1 | 8 | 898 | |||
XM_017011399.1 | XP_016866888.1 | 8 | 912 | |||
GenBank mRNA | AJ419865.1 | - | 9 | 951 | ||
AJ419866.1 | - | 9 | 1671 | |||
AK124363.1 | - | 2 | 2029 | |||
AK315467.1 | - | 9 | 958 | |||
BC001660.2 | - | 9 | 923 | 2959583 | ||
BC001819.2 | - | 9 | 923 | 2959583 | ||
BC036282.1 | - | 9 | 984 | |||
BC039713.1 | - | 9 | 926 | 5225327 | ||
BC051912.1 | - | 9 | 936 | 5227463 | ||
U85625.2 | - | 9 | 954 |