Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134885.1 | NP_001128357.1 | 11 | 1547 | ||
XM_008767258.1 | XP_008765480.1 | 11 | 2113 | |||
XM_008767259.1 | XP_008765481.1 | 11 | 2008 | |||
XM_008767260.1 | XP_008765482.1 | 11 | 2003 | |||
XM_008767261.1 | XP_008765483.1 | 10 | 2469 | |||
GenBank mRNA | FQ232022.1 | - | 4 | 602 | ||
FQ232863.1 | - | 2 | 310 |
Gene Symbol: LOC100911860 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767354.1 | XP_008765576.1 | 10 | 2469 |