Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108232.2 | NP_001101702.1 | 4 | 1556 | ||
XM_006245409.2 | XP_006245471.1 | 4 | 1066 | |||
XM_006245410.2 | XP_006245472.1 | 4 | 1071 | |||
GenBank mRNA | BC168978.1 | - | 4 | 1556 | 7455775 |