Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sned1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167842.1 | NP_001161314.1 | 1, 2 | 364 | ||
XM_006245526.2 | XP_006245588.1 | 1, 2 | 392 | |||
XM_008767373.1 | XP_008765595.1 | 1, 2 | 393 | |||
XM_008767374.1 | XP_008765596.1 | 1, 2 | 393 |