Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127657.2 | NP_001121129.2 | 2 | 660 | ||
GenBank mRNA | BC158655.1 | - | 2 | 660 | 8369055 |
Gene Symbol: LOC100909485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752019.3 | XP_003752067.1 | 2 | 694 |