Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103694587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773722.1 | XP_008771944.1 | 1 | 117 |
Gene Symbol: LOC103690902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773581.1 | XP_008771803.1 | 1 | 117 |
Gene Symbol: LOC103690904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773582.1 | XP_008771804.1 | 1 | 117 |
Gene Symbol: LOC103694516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773552.1 | XP_008771774.1 | 1 | 117 |
Gene Symbol: LOC681067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257516.2 | XP_006257578.1 | 1 | 326 |
Gene Symbol: LOC691030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773536.1 | XP_008771758.1 | 1 | 255 |
Gene Symbol: RGD1565430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257538.2 | XP_006257600.1 | 262 | |||
XM_008773538.1 | XP_008771760.1 | 262 |