Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ste2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007718.2 | NP_001007719.1 | 4 | 413 | ||
XM_006250793.2 | XP_006250855.1 | 5 | 704 | |||
XM_008770039.1 | XP_008768261.1 | 6 | 618 | |||
GenBank mRNA | BC088157.1 | - | 4 | 413 | 7323209 | |
S76490.1 | - | 4 | 336 | |||
U50205.1 | - | 3 | 306 |
Gene Symbol: Sult1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012883.1 | NP_037015.1 | 4 | 437 | ||
GenBank mRNA | M86758.1 | - | 4 | 437 | ||
S76489.1 | - | 4 | 336 | |||
U50204.1 | - | 3 | 306 |