Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108267.1 | NP_001101737.1 | 6 | 969 | ||
XM_006245818.2 | XP_006245880.1 | 6 | 1725 | |||
XM_006245819.2 | XP_006245881.1 | 6 | 1424 | |||
XM_006245820.2 | XP_006245882.1 | 6 | 1861 | |||
XM_006245822.2 | XP_006245884.1 | 6 | 2551 | |||
XM_006245823.2 | XP_006245885.1 | 6 | 1385 |