Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108288.1 | NP_001101758.1 | 15 | 1566 | ||
XM_006247099.2 | XP_006247161.1 | 14 | 1652 | |||
XM_006247100.2 | XP_006247162.1 | 14 | 1651 | |||
XM_006247101.2 | XP_006247163.1 | 14 | 1652 | |||
XM_006247102.2 | XP_006247164.1 | 14 | 1651 | |||
XM_006247104.2 | XP_006247166.1 | 14 | 1651 | |||
XM_006247105.2 | XP_006247167.1 | 14 | 1651 | |||
XM_008768074.1 | XP_008766296.1 | 14 | 1651 | |||
GenBank mRNA | BC168955.1 | - | 14 | 1487 | 7135665 |