Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126.3 | NP_001117.2 | 11 | 1400 | ||
XM_011544108.2 | XP_011542410.1 | 11 | 2072 | |||
GenBank mRNA | AK025514.1 | - | 11 | 1147 | ||
AK303886.1 | - | 12 | 1112 | |||
BC012356.1 | - | 11 | 1185 | 4635347 |