Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003786.3 | NP_003777.2 | 20 | 2749 | ||
XM_005257763.3 | XP_005257820.1 | 19 | 2571 | |||
XM_011525422.2 | XP_011523724.1 | 19 | 2676 | |||
XM_011525423.1 | XP_011523725.1 | 20 | 2868 | |||
XM_011525424.2 | XP_011523726.1 | 15 | 2099 | |||
XM_011525425.2 | XP_011523727.1 | 19 | 2576 | |||
XM_017025265.1 | XP_016880754.1 | 16 | 2612 | |||
XM_017025266.1 | XP_016880755.1 | 20 | 2868 | |||
GenBank mRNA | AB010887.1 | - | 20 | 2683 | ||
AB208954.1 | - | 20 | 2689 | |||
AF009670.1 | - | 20 | 2742 | |||
AF083552.1 | - | 20 | 2686 | |||
AF085690.1 | - | 20 | 2705 | |||
AF085691.1 | - | 20 | 2705 | |||
AF085692.1 | - | 20 | 2760 | |||
AF104943.1 | - | 20 | 2673 | |||
BC137347.1 | - | 20 | 2700 | 9021345 | ||
BC137348.1 | - | 20 | 2700 | 9021346 | ||
Y17151.2 | - | 20 | 2711 |