Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003794.3 | NP_003785.1 | 3 | 394 | ||
NR_073435.1 | 2 | 272 | ||||
XM_017007414.1 | XP_016862903.1 | 3 | 386 | |||
GenBank mRNA | AF065485.1 | - | 3 | 352 | ||
AK001835.1 | - | 3 | 356 | |||
AK298972.1 | - | 2 | 239 | |||
BC018762.1 | - | 3 | 370 | 4641014 | ||
BI669801.1 | - | 3 | 398 | |||
DA289870.1 | - | 3 | 396 |