Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108335.1 | NP_001101805.1 | 12 | 1291 | ||
XM_006249395.2 | XP_006249457.1 | 11 | 2488 | |||
XM_006249396.2 | XP_006249458.1 | 11 | 2492 |