Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077589.1 | NP_001071057.1 | 2 | 87 | ||
XM_006250010.2 | XP_006250072.1 | 3 | 998 | |||
XM_006250011.2 | XP_006250073.1 | 3 | 1003 | |||
XM_006250012.1 | XP_006250074.1 | 3 | 1064 | |||
GenBank mRNA | AY651775.1 | - | 2 | 87 |