Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003801.3 | NP_003792.1 | 2 | 257 | ||
GenBank mRNA | AB002135.1 | - | 2 | 242 | ||
AB006969.1 | - | 2 | 155 | |||
AK312759.1 | - | 2 | 269 | |||
AL157437.1 | - | 2 | 541 | |||
BC003171.1 | - | 2 | 233 | 2906111 | ||
BC004129.2 | - | 2 | 257 | 3504366 |