Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108365.1 | NP_001101835.1 | 3, 4 | 998 | ||
XM_006251340.2 | XP_006251402.1 | 2, 3 | 1033 | |||
XM_008770332.1 | XP_008768554.1 | 1, 2 | 1359 | |||
XM_008770333.1 | XP_008768555.1 | 1, 2 | 1358 |