Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ogdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017461.1 | NP_001017461.1 | 22, 23 | 3022 | ||
XM_006251446.2 | XP_006251508.1 | 22, 23 | 3138 | |||
XM_006251447.2 | XP_006251509.1 | 22, 23 | 3284 | |||
XM_006251448.2 | XP_006251510.1 | 22, 23 | 3061 | |||
GenBank mRNA | BC083811.1 | - | 22, 23 | 3022 | 7110182 | |
FQ222842.1 | - | 22, 23 | 2997 |
Gene Symbol: LOC103693780 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770414.1 | XP_008768636.1 | 1924 |