Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgalsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134730.1 | NP_001128202.1 | 4 | 403 | ||
GenBank mRNA | BC168193.1 | - | 4 | 403 | 7109522 | |
FQ211679.1 | - | 4 | 247 |